4-[5-(Trifluoromethyl)pyridin-2-yl]benzyl alcohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[4-(Hydroxymethyl)phenyl]-5-(trifluoromethyl)pyridine, {4-[5-(Trifluoromethyl)pyridin-2-yl]phenyl}methanol
PC8553-1G
PC8553-250MG
APOSPC8553-1GEA
343
EUR
APOSPC8553-1G
APOSPC8553-250MG
4-[5-(Trifluoromethyl)pyridin-2-yl]benzyl alcohol 97%
4-[5-(Trifluoromethyl)pyridin-2-yl]benzyl alcohol
Formel:
C₁₃H₁₀F₃NO Molecular Weight: 253.22 g/mol Schmelzpunkt: 123…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817477 CAS-Nummer: 613239-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...