5-Nitro-2,3,4-trifluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,4-Trifluoro-5-nitrobenzoic acid

PC8688-25G PC8688-5G
APOSPC8688-25GEA 121 EUR
APOSPC8688-25G APOSPC8688-5G
5-Nitro-2,3,4-trifluorobenzoic acid
5-Nitro-2,3,4-trifluorobenzoic acid
Formel: C₇H₂F₃NO₄
Molecular Weight: 221.09 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09753762
CAS-Nummer: 197520-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR