2-Bromo-1-fluoronaphthalene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC8694-1G
PC8694-5G
APOSPC8694-1GEA
139
EUR
APOSPC8694-1G
APOSPC8694-5G
2-Bromo-1-fluoronaphthalene 95%
2-Bromo-1-fluoronaphthalene
Formel:
C₁₀H₆BrF Molecular Weight: 225,057 g/mol Siedepunkt: 164 °C (2 mm Hg) Schmelzpunkt: –4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864690 CAS-Nummer: 317-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...