Perfluorophenyl pyridine-2-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pentafluorophenyl picolinate, Pentafluorophenyl pyridine-2-carboxylate, 2-[(Pentafluorophenoxy)carbonyl]pyridine
PC8772-1G
PC8772-250MG
APOSPC8772-1GEA
285
EUR
APOSPC8772-1G
APOSPC8772-250MG
Perfluorophenyl pyridine-2-carboxylate 95%
Perfluorophenyl pyridine-2-carboxylate
Formel:
C₁₂H₄F₅NO₂ Molecular Weight: 289,158 g/mol Schmelzpunkt: 55,5…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02916457 CAS-Nummer: 188837-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...