1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC8773-1G PC8773-5G
APOSPC8773-1GEA 69.6 EUR
APOSPC8773-1G APOSPC8773-5G
1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carboxaldehyde 97%
1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carboxaldehyde
Formel: C₁₂H₈F₃NO
Molecular Weight: 239.193 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01567130
CAS-Nummer: 156496-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR