1-(2-Chloro-6-fluorobenzyl)homopiperazine 97%

Lieferant: Apollo Scientific
Gefahr

PC8800-1G PC8800-5G PC8800-10G
APOSPC8800-1GEA 149 EUR
APOSPC8800-1G APOSPC8800-5G APOSPC8800-10G
1-(2-Chloro-6-fluorobenzyl)homopiperazine 97%
1-(2-Chloro-6-fluorobenzyl)homopiperazine
Formel: C₁₂H₁₆ClFN₂
Molecular Weight: 242,72 g/mol
Siedepunkt: 103 °C (0,1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00816470
CAS-Nummer: 244022-69-3
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR