2-[4-(Trifluoromethyl)phenyl]pyrrolidine 98%

Lieferant: Apollo Scientific
Achtung

PC8823-1G PC8823-250MG
APOSPC8823-1GEA 256 EUR
APOSPC8823-1G APOSPC8823-250MG
2-[4-(Trifluoromethyl)phenyl]pyrrolidine 98%
2-[4-(Trifluoromethyl)phenyl]pyrrolidine
Formel: C₁₁H₁₂F₃N
Molecular Weight: 215,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631837
CAS-Nummer: 298690-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR