4-(Aminomethyl)-3-fluoropyridine dihydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Fluoroisonicotinylamine dihydrochloride, (3-Fluoropyridin-4-yl)methylamine dihydrochloride, (3-Fluoropyridin-4-yl)methanamine dihydrochloride

PC8935-250MG PC8935-500MG
APOSPC8935-250MGEA 418 EUR
APOSPC8935-250MG APOSPC8935-500MG
4-(Aminomethyl)-3-fluoropyridine dihydrochloride
4-(Aminomethyl)-3-fluoropyridine dihydrochloride
Formel: C₆H₇FN₂·2ClH
Molecular Weight: 199,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12922842
CAS-Nummer: 1257535-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR