2-(Aminomethyl)-4-fluoropyridine dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluoropyridin-2-yl)methylamine dihydrochloride, 4-Fluoropicolinylamine dihydrochloride
PC8936-250MG
PC8936-100MG
APOSPC8936-250MGEA
275
EUR
APOSPC8936-250MG
APOSPC8936-100MG
2-(Aminomethyl)-4-fluoropyridine dihydrochloride
2-(Aminomethyl)-4-fluoropyridine dihydrochloride
Formel:
C₆H₇FN₂·2ClH Molecular Weight: 199,053 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12922841 CAS-Nummer: 1257535-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...