3-Fluoropropylamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Amino-3-fluoropropane hydrochloride

PC8996-1G PC8996-250MG
APOSPC8996-1GEA 406 EUR
APOSPC8996-1G APOSPC8996-250MG
3-Fluoropropylamine hydrochloride 97%
3-Fluoropropylamine hydrochloride
Formel: C₃H₈FN·ClH
Molecular Weight: 113,562 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07369956
CAS-Nummer: 64068-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR