1-(5-Bromo-2-fluorophenyl)propan-1-one 96%

Lieferant: Apollo Scientific
Achtung

PC900041-1G PC900041-5G
APOSPC900041-1GEA 190 EUR
APOSPC900041-1G APOSPC900041-5G
1-(5-Bromo-2-fluorophenyl)propan-1-one 96%
1-(5-Bromo-2-fluorophenyl)propan-1-one
Formel: C₉H₈BrFO
Molecular Weight: 231,062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226832
CAS-Nummer: 864774-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR