4-(3-Fluorophenoxy)aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(3-Fluorophenoxy)aniline
PC900144-1G
PC900144-5G
APOSPC900144-1GEA
105
EUR
APOSPC900144-1G
APOSPC900144-5G
4-(3-Fluorophenoxy)aniline
4-(3-Fluorophenoxy)aniline
Formel:
C₁₂H₁₀FNO Molecular Weight: 203,212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663349 CAS-Nummer: 307308-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...