1-(2-Fluoro-5-hydroxyphenyl)ethanone 98%

Lieferant: Apollo Scientific

PC900169-1G PC900169-5G
APOSPC900169-1GEA 91.5 EUR
APOSPC900169-1G APOSPC900169-5G
1-(2-Fluoro-5-hydroxyphenyl)ethanone 98%
1-(2-Fluoro-5-hydroxyphenyl)ethanone
Formel: C₈H₇FO₂
Molecular Weight: 154.138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038472
CAS-Nummer: 145300-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR