4-Fluoro-2-[(1E)-(hydroxyimino)methyl]phenol 97%
Lieferant: Apollo Scientific
PC900171-1G
PC900171-5G
APOSPC900171-1GEA
132
EUR
APOSPC900171-1G
APOSPC900171-5G
4-Fluoro-2-[(1E)-(hydroxyimino)methyl]phenol 97%
4-Fluoro-2-[(1E)-(hydroxyimino)methyl]phenol
Formel:
C₇H₆FNO₂ Molecular Weight: 155,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442128 CAS-Nummer: 91407-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...