[2-Amino-1-(2-fluorophenyl)ethyl]dimethylamine 95%

Lieferant: Apollo Scientific

PC900193-1G PC900193-250MG
APOSPC900193-1GEA 502 EUR
APOSPC900193-1G APOSPC900193-250MG
[2-Amino-1-(2-fluorophenyl)ethyl]dimethylamine 95%
[2-Amino-1-(2-fluorophenyl)ethyl]dimethylamine
Formel: C₁₀H₁₅FN₂
Molecular Weight: 182.238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06753110
CAS-Nummer: 842954-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR