5-Fluoro-2-(2-methoxyethoxy)pyridine 98%

Lieferant: Apollo Scientific

PC900589-1G PC900589-5G
APOSPC900589-1GEA 232 EUR
APOSPC900589-1G APOSPC900589-5G
5-Fluoro-2-(2-methoxyethoxy)pyridine 98%
5-Fluoro-2-(2-methoxyethoxy)pyridine
Formel: C₈H₁₀FNO₂
Molecular Weight: 171,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18914536
CAS-Nummer: 1305322-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR