6-Chloro-5-fluoropyridine-3-carbaldehyde 97%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-5-fluoronicotinaldehyde, 2-Chloro-3-fluoropyridine-3-carboxaldehyde

PC900606-250MG
APOSPC900606-250MGEA 91.5 EUR
APOSPC900606-250MG
6-Chloro-5-fluoropyridine-3-carbaldehyde 97%
6-Chloro-5-fluoropyridine-3-carbaldehyde
Formel: C₆H₃ClFNO
Molecular Weight: 159,546 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189060
CAS-Nummer: 950691-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR