5-Bromo-4-methyl-2-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

PC900609-100MG PC900609-1G PC900609-250MG
APOSPC900609-100MGEA 95.7 EUR
APOSPC900609-100MG APOSPC900609-1G APOSPC900609-250MG
5-Bromo-4-methyl-2-(trifluoromethyl)pyridine 95%
5-Bromo-4-methyl-2-(trifluoromethyl)pyridine
Formel: C₇H₅BrF₃N
Molecular Weight: 240.021 g/mol
Siedepunkt: 203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18257679
CAS-Nummer: 1010422-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR