8-Fluoro-2-methylquinoline 95%

Lieferant: Apollo Scientific

PC900629-100MG PC900629-1G PC900629-250MG
APOSPC900629-100MGEA 0 EUR
APOSPC900629-100MG APOSPC900629-1G APOSPC900629-250MG
8-Fluoro-2-methylquinoline 95%
8-Fluoro-2-methylquinoline
Formel: C₁₀H₈FN
Lagertemperatur: Raumtemperatur
MDL: MFCD09744004
CAS-Nummer: 46001-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR