2-(4-Fluoronaphthalen-1-yl)acetic acid 95%

Lieferant: Apollo Scientific

PC901232-1G PC901232-250MG
APOSPC901232-1GEA 647 EUR
APOSPC901232-1G APOSPC901232-250MG
2-(4-Fluoronaphthalen-1-yl)acetic acid 95%
2-(4-Fluoronaphthalen-1-yl)acetic acid
Formel: C₁₂H₉FO₂
Molecular Weight: 204,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832460
CAS-Nummer: 3833-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR