6-Ethoxy-2-fluoropyridine-3-boronic acid 98%

Lieferant: Apollo Scientific

PC901346-1G PC901346-250MG
APOSPC901346-1GEA 574 EUR
APOSPC901346-1G APOSPC901346-250MG
6-Ethoxy-2-fluoropyridine-3-boronic acid 98%
6-Ethoxy-2-fluoropyridine-3-boronic acid
Formel: C₇H₉BFNO₃
Molecular Weight: 184,961 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18434410
CAS-Nummer: 2096335-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR