(5-chloro-6-fluoropyridin-3-yl)boronic acid 96%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2-fluoropyridne-5-boronic acid, (5-Chloro-6-fluoropyridin-3-yl)boronic acid

PC901355-1G PC901355-250MG
APOSPC901355-1GEA 201 EUR
APOSPC901355-1G APOSPC901355-250MG
(5-chloro-6-fluoropyridin-3-yl)boronic acid 96%
(5-chloro-6-fluoropyridin-3-yl)boronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175,353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18257900
CAS-Nummer: 1366482-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR