2-Fluoro-3-(methoxymethoxy)phenylboronic acid 98%
Lieferant: Apollo Scientific
PC901369-1G
PC901369-5G
APOSPC901369-1GEA
219
EUR
APOSPC901369-1G
APOSPC901369-5G
2-Fluoro-3-(methoxymethoxy)phenylboronic acid 98%
2-Fluoro-3-(methoxymethoxy)phenylboronic acid
Formel:
C₈H₁₀BFO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18914499 CAS-Nummer: 2096331-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...