4-Fluoro-3-formyl-5-methylphenylboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (4-Fluoro-3-formyl-5-methylphenyl)boronic acid

PC901441-1G PC901441-5G
APOSPC901441-1GEA 82.8 EUR
APOSPC901441-1G APOSPC901441-5G
4-Fluoro-3-formyl-5-methylphenylboronic acid 98%
4-Fluoro-3-formyl-5-methylphenylboronic acid
Formel: C₈H₈BFO₃
Molecular Weight: 181,957 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111909
CAS-Nummer: 1310384-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR