Methyl 3-(4-fluorophenyl)propanoate 98%

Lieferant: Apollo Scientific

PC901528-1G PC901528-5G
APOSPC901528-1GEA 179 EUR
APOSPC901528-1G APOSPC901528-5G
Methyl 3-(4-fluorophenyl)propanoate 98%
Methyl 3-(4-fluorophenyl)propanoate
Formel: C₁₀H₁₁FO₂
Molecular Weight: 182.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031748
CAS-Nummer: 2928-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR