2-Ethoxy-3-fluoropyridine-5-boronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
(6-Ethoxy-5-fluoropyridin-3-yl)boronic acid
PC901636-1G
PC901636-250MG
APOSPC901636-1GEA
734
EUR
APOSPC901636-1G
APOSPC901636-250MG
2-Ethoxy-3-fluoropyridine-5-boronic acid 95%
2-Ethoxy-3-fluoropyridine-5-boronic acid
Formel:
C₇H₉BFNO₃ Molecular Weight: 184,961 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07374965 CAS-Nummer: 1309982-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...