2,2,2-Trifluoro-1-(3-hydroxypiperidin-1-yl)ethanone 96%

Lieferant: Apollo Scientific

PC901648-1G PC901648-250MG
APOSPC901648-1GEA 806 EUR
APOSPC901648-1G APOSPC901648-250MG
2,2,2-Trifluoro-1-(3-hydroxypiperidin-1-yl)ethanone 96%
2,2,2-Trifluoro-1-(3-hydroxypiperidin-1-yl)ethanone
Formel: C₇H₁₀F₃NO₂
Molecular Weight: 197,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12777054
CAS-Nummer: 73193-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR