6-Trifluoromethylquinoline 98%

Lieferant: Apollo Scientific

Synonyme: 6-(Trifluoromethyl)quinoline

PC901832-1G PC901832-5G
APOSPC901832-1GEA 97.3 EUR
APOSPC901832-1G APOSPC901832-5G
6-Trifluoromethylquinoline 98%
6-Trifluoromethylquinoline
Formel: C₁₀H₆F₃N
Molecular Weight: 197,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09879698
CAS-Nummer: 325-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR