4-Bromo-1-ethyl-3-(trifluoromethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC901838-1G PC901838-5G
APOSPC901838-1GEA 34.8 EUR
APOSPC901838-1G APOSPC901838-5G
4-Bromo-1-ethyl-3-(trifluoromethyl)-1H-pyrazole
4-Bromo-1-ethyl-3-(trifluoromethyl)-1H-pyrazole
Formel: C₆H₆BrF₃N₂
Molecular Weight: 243,024 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976413
CAS-Nummer: 1245772-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR