Difluoro(4-fluorophenyl)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2,2-Difluoro-2-(4-fluorophenyl)acetic Acid

PC901906-1G PC901906-5G
APOSPC901906-1GEA 106 EUR
APOSPC901906-1G APOSPC901906-5G
Difluoro(4-fluorophenyl)acetic acid 95%
Difluoro(4-fluorophenyl)acetic acid
Formel: C₈H₅F₃O₂
Molecular Weight: 190,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11167795
CAS-Nummer: 94010-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR