6-Fluoro-8-methylquinoline 98%

Lieferant: Apollo Scientific

PC901909-1G PC901909-5G
APOSPC901909-1GEA 219 EUR
APOSPC901909-1G APOSPC901909-5G
6-Fluoro-8-methylquinoline 98%
6-Fluoro-8-methylquinoline
Formel: C₁₀H₈FN
Molecular Weight: 161,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026038
CAS-Nummer: 1150271-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR