4,6-Difluorobenzo[c][1,2,5]thiadiazole 95%

Lieferant: Apollo Scientific

PC901965-1G PC901965-5G
APOSPC901965-1GEA 248 EUR
APOSPC901965-1G APOSPC901965-5G
4,6-Difluorobenzo[c][1,2,5]thiadiazole 95%
4,6-Difluorobenzo[c][1,2,5]thiadiazole
Formel: C₆H₂F₂N₂S
Molecular Weight: 172.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036121
CAS-Nummer: 1221793-59-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR