6-Bromo-5-fluoro-1H-benzo[d][1,2,3]triazole 95+%

Lieferant: Apollo Scientific
Achtung

PC901978-1G PC901978-250MG PC901978-5G
APOSPC901978-1GEA 75.4 EUR
APOSPC901978-1G APOSPC901978-250MG APOSPC901978-5G
6-Bromo-5-fluoro-1H-benzo[d][1,2,3]triazole 95+%
6-Bromo-5-fluoro-1H-benzo[d][1,2,3]triazole
Formel: C₆H₃BrFN₃
Molecular Weight: 216,011 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16660325
CAS-Nummer: 1242336-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR