2,3-Difluoro-6-nitrotoluene 95%

Lieferant: Apollo Scientific

PC902026-10G PC902026-1G PC902026-5G
APOSPC902026-10GEA 300 EUR
APOSPC902026-10G APOSPC902026-1G APOSPC902026-5G
2,3-Difluoro-6-nitrotoluene 95%
2,3-Difluoro-6-nitrotoluene
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08706407
CAS-Nummer: 914348-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR