2-(2-Fluorophenyl)-3-oxobutanenitrile 97%

Lieferant: Apollo Scientific

PC902061-1G PC902061-5G
APOSPC902061-1GEA 219 EUR
APOSPC902061-1G APOSPC902061-5G
2-(2-Fluorophenyl)-3-oxobutanenitrile 97%
2-(2-Fluorophenyl)-3-oxobutanenitrile
Formel: C₁₀H₈FNO
Molecular Weight: 177,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00055279
CAS-Nummer: 93839-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR