N-[(3-fluorophenyl)methyl]cyclopropanamine 98%
Lieferant: Apollo Scientific
PC902078-1G
PC902078-5G
APOSPC902078-1GEA
219
EUR
APOSPC902078-1G
APOSPC902078-5G
N-[(3-fluorophenyl)methyl]cyclopropanamine 98%
N-[(3-fluorophenyl)methyl]cyclopropanamine
Formel:
C₁₀H₁₂FN Molecular Weight: 165,207 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08060645 CAS-Nummer: 920479-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...