2-Methyl-5-(trifluoromethyl)pyrazole-3-carbaldehyde 97%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbaldehyde

PC902089-1G PC902089-250MG
APOSPC902089-1GEA 343 EUR
APOSPC902089-1G APOSPC902089-250MG
2-Methyl-5-(trifluoromethyl)pyrazole-3-carbaldehyde 97%
2-Methyl-5-(trifluoromethyl)pyrazole-3-carbaldehyde
Formel: C₆H₅F₃N₂O
Molecular Weight: 178,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22398340
CAS-Nummer: 1414962-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR