1-Ethyl-3-(trifluoromethyl)pyrazole-4-carbaldehyde 98%
Lieferant: Apollo Scientific
PC902099-1G
PC902099-250MG
APOSPC902099-1GEA
676
EUR
APOSPC902099-1G
APOSPC902099-250MG
1-Ethyl-3-(trifluoromethyl)pyrazole-4-carbaldehyde 98%
1-Ethyl-3-(trifluoromethyl)pyrazole-4-carbaldehyde
Formel:
C₇H₇F₃N₂O Molecular Weight: 192.138 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25949488 CAS-Nummer: 1443279-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...