4-Fluoro-N-methylbenzylamine hydrochloride 97%

Lieferant: Apollo Scientific

PC902106-10G PC902106-25G
APOSPC902106-10GEA 190 EUR
APOSPC902106-10G APOSPC902106-25G
4-Fluoro-N-methylbenzylamine hydrochloride 97%
4-Fluoro-N-methylbenzylamine hydrochloride
Formel: C₈H₁₀FN·ClH
Molecular Weight: 175,631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07106763
CAS-Nummer: 459-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR