2,6-Difluoro-N,N-dimethylbenzamide 98%

Lieferant: Apollo Scientific

PC902116-1G PC902116-5G
APOSPC902116-1GEA 168 EUR
APOSPC902116-1G APOSPC902116-5G
2,6-Difluoro-N,N-dimethylbenzamide 98%
2,6-Difluoro-N,N-dimethylbenzamide
Formel: C₉H₉F₂NO
Molecular Weight: 185,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20356572
CAS-Nummer: 81652-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR