Methyl 3,5-difluoro-4-hydroxybenzoate 98%
Lieferant: Apollo Scientific
PC902120-10G
PC902120-1G
PC902120-5G
APOSPC902120-10GEA
964
EUR
APOSPC902120-10G
APOSPC902120-1G
APOSPC902120-5G
Methyl 3,5-difluoro-4-hydroxybenzoate 98%
Methyl 3,5-difluoro-4-hydroxybenzoate
Formel:
C₈H₆F₂O₃ Molecular Weight: 188.128 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12406862 CAS-Nummer: 170572-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...