3-Fluoro-5-nitroanisole 97%

Lieferant: Apollo Scientific

Synonyme: 1-Fluoro-3-methoxy-5-nitrobenzene

PC902146-10G PC902146-1G PC902146-5G
APOSPC902146-10GEA 690 EUR
APOSPC902146-10G APOSPC902146-1G APOSPC902146-5G
3-Fluoro-5-nitroanisole 97%
3-Fluoro-5-nitroanisole
Formel: C₇H₆FNO₃
Molecular Weight: 171.126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848484
CAS-Nummer: 7087-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR