3-(4-Fluorophenyl)-5-methyl-1,2,4-oxadiazole 98%

Lieferant: Apollo Scientific

PC902203-1G PC902203-25G PC902203-5G
APOSPC902203-1GEA 145 EUR
APOSPC902203-1G APOSPC902203-25G APOSPC902203-5G
3-(4-Fluorophenyl)-5-methyl-1,2,4-oxadiazole 98%
3-(4-Fluorophenyl)-5-methyl-1,2,4-oxadiazole
Formel: C₉H₇FN₂O
Molecular Weight: 178,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17214258
CAS-Nummer: 196301-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR