4-Chloro-2-fluoro-5-nitroaniline 98%

Lieferant: Apollo Scientific

PC902215-1G PC902215-5G
APOSPC902215-1GEA 270 EUR
APOSPC902215-1G APOSPC902215-5G
4-Chloro-2-fluoro-5-nitroaniline 98%
4-Chloro-2-fluoro-5-nitroaniline
Formel: C₆H₄ClFN₂O₂
Molecular Weight: 190,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04037232
CAS-Nummer: 86988-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR