5-Fluoro-1-methylbenzimidazole 98%
Lieferant: Apollo Scientific
PC902269-1G
PC902269-250MG
PC902269-5G
APOSPC902269-1GEA
270
EUR
APOSPC902269-1G
APOSPC902269-250MG
APOSPC902269-5G
5-Fluoro-1-methylbenzimidazole 98%
5-Fluoro-1-methylbenzimidazole
Formel:
C₈H₇FN₂ Molecular Weight: 150.153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18378522 CAS-Nummer: 1365271-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...