6,7-Difluoro-1,2-dimethyl-1,3-benzodiazole 98%

Lieferant: Apollo Scientific

PC902286-1G PC902286-5G PC902286-10G
APOSPC902286-1GEA 145 EUR
APOSPC902286-1G APOSPC902286-5G APOSPC902286-10G
6,7-Difluoro-1,2-dimethyl-1,3-benzodiazole 98%
6,7-Difluoro-1,2-dimethyl-1,3-benzodiazole
Formel: C₉H₈F₂N₂
Molecular Weight: 182.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22375039
CAS-Nummer: 1381944-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR