5-Fluoro-N-isopropyl-2-nitroaniline 98%

Lieferant: Apollo Scientific

PC902319-10G PC902319-1G PC902319-5G
APOSPC902319-10GEA 502 EUR
APOSPC902319-10G APOSPC902319-1G APOSPC902319-5G
5-Fluoro-N-isopropyl-2-nitroaniline 98%
5-Fluoro-N-isopropyl-2-nitroaniline
Formel: C₉H₁₁FN₂O₂
Molecular Weight: 198,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16744202
CAS-Nummer: 131885-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR