2-(4-Fluorophenyl)benzonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 4'-Fluoro-[1,1'-biphenyl]-2-carbonitrile

PC902362-1G PC902362-5G
APOSPC902362-1GEA 145 EUR
APOSPC902362-1G APOSPC902362-5G
2-(4-Fluorophenyl)benzonitrile 98%
2-(4-Fluorophenyl)benzonitrile
Formel: C₁₃H₈FN
Molecular Weight: 197,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05979720
CAS-Nummer: 89346-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR