3-(4-Fluorophenyl)benzonitrile 98%

Lieferant: Apollo Scientific

PC902446-1G PC902446-5G
APOSPC902446-1GEA 219 EUR
APOSPC902446-1G APOSPC902446-5G
3-(4-Fluorophenyl)benzonitrile 98%
3-(4-Fluorophenyl)benzonitrile
Formel: C₁₃H₈FN
Molecular Weight: 197,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05979962
CAS-Nummer: 10540-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR