5-Fluoro-4-methyl-2-nitroanisole 98%

Lieferant: Apollo Scientific

Synonyme: 1-Fluoro-5-methoxy-2-methyl-4-nitrobenzene

PC902481-1G PC902481-250MG
APOSPC902481-1GEA 91.5 EUR
APOSPC902481-1G APOSPC902481-250MG
5-Fluoro-4-methyl-2-nitroanisole 98%
5-Fluoro-4-methyl-2-nitroanisole
Formel: C₈H₈FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15527524
CAS-Nummer: 314298-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR